missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SLFN5 (aa 70-210) Control Fragment Recombinant Protein

Catalog No. RP90825
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP90825 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP90825 Supplier Invitrogen™ Supplier No. RP90825
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (51%), Rat (51%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53638 (PA5-53638. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

SLFN5 may have a role in hematopoietic cell differentiation.

Specifications

Accession Number Q08AF3
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 162394
Name Human SLFN5 (aa 70-210) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias Schlafen family member 5; SLFN5
Common Name SLFN5
Gene Symbol SLFN5
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ERHGVGLDVPPIFRSHLDKMQKENHFLIFVKSWNTEAGVPLATLCSNLYHRERTSTDVMDSQEALAFLKCRTQTPTNINVSNSLGPQAAQGSVQYEGNINVSAAALFDRKRLQYLEKLNLPESTHVEFVMFSTDVSHCVKD
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less