missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SKIP Synthetic Peptide

Catalog No. PEP0402
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0402 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0402 Supplier Invitrogen™ Supplier No. PEP0402
Only null left

Synthetic Peptide

SkiP Synthetic Peptide, PEP-0402, from Invitrogen.

This gene encodes a protein with 5-phosphatase activity toward polyphosphate inositol. The protein localizes to the cytosol in regions lacking actin stress fibers. It is thought that this protein may negatively regulate the actin cytoskeleton. Alternatively spliced transcript variants encoding different isoforms have been identified.

Specifications

Gene ID (Entrez) 51763
Species Human
Purification Method purified
Accession Number Q9BT40
Common Name Human SKIP
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol INPP5K
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less