missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SH2D4B (aa 170-239) Control Fragment Recombinant Protein

Catalog No. RP96525
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP96525 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP96525 Supplier Invitrogen™ Supplier No. RP96525
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-59032 (PA5-59032. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

SH2D4B (SH2 Domain Containing 4B) is a Protein Coding gene. Diseases associated with SH2D4B includeLymphocytic Vasculitis.

Specifications

Accession Number Q5SQS7
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 387694
Name Human SH2D4B (aa 170-239) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias novel SH2 domain-containing protein; SH2 domain containing 4 B; SH2 domain-containing protein 4 B; SH2D4B
Common Name SH2D4B
Gene Symbol SH2D4B
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ERKRGEEQIRLQEEQRAKELYWTLKQAQLHCQASEKEEREWEEQLRRSKAADEERSRRAQRARDEYRHHS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less