missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SAMD9L (aa 1-93) Control Fragment Recombinant Protein

Catalog No. RP92804
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP92804 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP92804 Supplier Invitrogen™ Supplier No. RP92804
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (70%), Rat (70%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53994 (PA5-53994. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The function of this protein has not been specifically defined.

Specifications

Accession Number Q8IVG5
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 219285
Name Human SAMD9L (aa 1-93) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AA175286; C7orf6; DRIF2; ESTM25; KIAA2005; mKIAA2005; SAM domain-containing protein 9-like; SAMD9L; sterile alpha motif domain containing 9 like; sterile alpha motif domain containing 9-like; sterile alpha motif domain-containing protein 9-like; UEF; UEF1; v-myc myelocytomatosis viral related oncogene, neuroblastoma derived
Common Name SAMD9L
Gene Symbol SAMD9L
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MSKQVSLPEMIKDWTKEHVKKWVNEDLKINEQYGQILLSEEVTGLVLQELTEKDLVEMGLPWGPALLIKRSYNKLNSKSPESDNHDPGQLDNS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less