missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human RANBP2 (aa 1-18) Synthetic Peptide

Catalog No. PEP137
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP137 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP137 Supplier Invitrogen™ Supplier No. PEP137

Synthetic Peptide

Human RanBP2 (reaidues 1-18) Synthetic Peptide, PEP-137, from Invitrogen.

RanBP2 interacts with the small GTP-binding protein Ran and forms a large multiprotein complex involved in many biological processes such as mRNA processing, nuclear transport, cell cycle control and postmitotic nuclear reassembly. The protein is identified by western blotting as an apparent molecular mass of 360 kD. It is involved in nucleation of microtubule and control of cell growth by interacting with many protein factors.

Specifications

Gene ID (Entrez) 5903
Species Human
Purification Method purified
Accession Number P49792
Common Name Human RANBP2 (aa 1-18)
Concentration 0.5 mg/mL
Content And Storage -20°C. Avoid freeze-thaw cycles.
Description Synthetic Peptide
Format Lyophilized
Formulation peptide with no preservative
For Use With (Application) Control
Gene Symbol RANBP2
Lyophilized Lyophilized
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Sequence M(1)-R-R-S-K-A-D-V-E-R-Y-I-A-S-V-Q-G-S(18)-C
Show More Show Less