Learn More
ACROBiosystems Inc Human PVRIG Protein, Mouse IgG2a Fc Tag (MALS verified)
Catalog No. 502054952
Encompass
Recombinant Protein;1MG;Human PVRIG, Mouse IgG2a Fc Tag (PVG-H5259) is expressed from human 293 cells (HEK293). It contains AA Thr 41 - Asp 171 (Accession # Q6DKI7-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 40.7 kDa. The protein migrates as 50-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Nectin-2, His Tag (Cat. No. PV2-H52E2) at 5 g/mL (100 L/well) can bind Human PVRIG, Mouse IgG2a Fc Tag (Cat. No. PVG-H5259) with a linear range of 2-39 ng/mL (QC tested).
Catalog No. 502054952 Supplier ACROBiosystems Inc Supplier No. PVGH52591MG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;1MG;Human PVRIG, Mouse IgG2a Fc Tag (PVG-H5259) is expressed from human 293 cells (HEK293). It contains AA Thr 41 - Asp 171 (Accession # Q6DKI7-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 40.7 kDa. The protein migrates as 50-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Nectin-2, His Tag (Cat. No. PV2-H52E2) at 5 g/mL (100 L/well) can bind Human PVRIG, Mouse IgG2a Fc Tag (Cat. No. PVG-H5259) with a linear range of 2-39 ng/mL (QC tested).