Learn More
Invitrogen™ Human PPAR gamma-2 (aa 1-16) Synthetic Peptide
Description
PEP-027 is a 16 amino acid synthetic peptide whose sequence is derived from human PPARg2 and is absent in PPARg1. A COOH-terminal cysteine residue was added for conjugation to KLH. The sequence of this peptide is (amino to carboxy terminus): M (1) G E T L G D S P I D P E S D S (16) C This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and PPARg2, catalog # PA1-824. Using a solution with equal weights per unit volume of peptide and corresponding antibody will yield a solution with a large molar excess of peptide that is able to competitively bind the antibody. Reconstitute with 0.1 mL of distilled water.
Specifications
Specifications
| Gene ID (Entrez) | 5468 |
| Species | Human |
| Purification Method | purified |
| Accession Number | P37231 |
| Common Name | PPAR gamma-2 |
| Concentration | 0.5 mg/mL |
| Content And Storage | -20°C. Avoid freeze-thaw cycles. |
| Description | Synthetic Peptide |
| Format | Lyophilized |
| Formulation | Peptide with no preservative |
| Show More |