missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PDE4C (aa 684-712) Control Fragment Recombinant Protein

Catalog No. RP108909
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP108909 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP108909 Supplier Invitrogen™ Supplier No. RP108909
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (41%), Rat (41%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Cyclic nucleotides are important second messengers that regulate and mediate a number of cellular responses to extracellular signals, such as hormones, light, and neurotransmitters. Cyclic nucleotide phosphodiesterases.

Specifications

Accession Number Q08493
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 5143
Name Human PDE4C (aa 684-712) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias cAMP-specific 3',5'-cyclic phosphodiesterase 4 C; cAMP-specific 35-cyclic phosphodiesterase 4 C; CN4C; DPDE1; dunce; E130301F19Rik; MGC126222; PDE21; Pde4c; phosphodiesterase 4 C; phosphodiesterase 4 C, cAMP specific; phosphodiesterase 4 C, cAMP-specific; phosphodiesterase 4 C, cAMP-specific (phosphodiesterase E1 dunce homolog, Drosophila); phosphodiesterase E1 dunce homolog
Common Name PDE4C
Gene Symbol PDE4C
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence EALELPDTELLSPEAGPDPGDLPLDNQRT
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less