missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human NRK (aa 849-968) Control Fragment Recombinant Protein

Catalog No. RP89780
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP89780 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP89780 Supplier Invitrogen™ Supplier No. RP89780
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (70%), Rat (70%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53566 (PA5-53566. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The mouse ortholog of this gene encodes a protein kinase required for JNK activation. The encoded protein may be involved in the induction of actin polymerization in late embryogenesis.

Specifications

Accession Number Q7Z2Y5
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 203447
Name Human NRK (aa 849-968) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AA407323; AA409510; AI225895; AV168563; AW538185; Nck-interacting kinase-like embryo specific kinase; Nesk; Nik related kinase; NIK-like embryo-specific kinase; nik-related protein kinase; Nrk
Common Name NRK
Gene Symbol NRK
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GRRSQSSPPYSTIDQKLLVDIHVPDGFKVGKISPPVYLTNEWVGYNALSEIFRNDWLTPAPVIQPPEEDGDYVELYDASADTDGDDDDESNDTFEDTYDHANGNDDLDNQVDQANDVCKD
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less