missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human NOXRED1 (aa 231-333) Control Fragment Recombinant Protein

Catalog No. RP99869
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP99869 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP99869 Supplier Invitrogen™ Supplier No. RP99869
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (64%), Rat (64%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63155 (PA5-63155. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Probable oxidoreductase.

Specifications

Accession Number Q6NXP6
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 122945
Name Human NOXRED1 (aa 231-333) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 4933437F05Rik; C14orf148; NADP dependent oxidoreductase domain containing 1; NADP+ dependent oxidoreductase domain containing 1; NADP-dependent oxidoreductase domain containing 1; NADP-dependent oxidoreductase domain-containing protein 1; Noxred1; Pyrroline-5-carboxylate reductase-like protein C14orf148; pyrroline-5-carboxylate reductase-like protein C14orf148 homolog
Common Name NOXRED1
Gene Symbol NOXRED1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ILNIKWLEGVFYAALNICTARNMAHSQVLQLLSELFLSVHFEDCGKDTASCPKLQLTDFVSKAYGKNLSQERPFPWFDLTAVQLKETPFSQHLSSSPVLQDHL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less