missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human NFAT5 (aa 1439-1456) Synthetic Peptide
Description
Human NFAT5 (residues 1439-1456) Synthetic Peptide, PEP-123, from Invitrogen.
The nuclear factor of activated T-cells (NFAT) transcription complex is required for the expression of a group of proteins that collectively regulate the immune response. Four NFAT proteins, encoded on separate genes and expressed as several splice variants, have been described: NFAT1 (also known as NFATp or NFATc2), NFAT2 (NFATc or NFATc1), NFAT3, and NFAT4 (NFATx or NFATc3). These proteins show a low level of sequence similarity with the Dorsal/Rel/NFkB family of transcription factors. Another NFAT-related protein termed NFAT5 differs from isoforms 1-4 in that it lacks many of the Fos/Jun contact sites observed in its predecessors and its subcellular localization is not calcineurin-dependent.
Specifications
Specifications
| Gene ID (Entrez) | 10725 |
| Species | Human |
| Purification Method | purified |
| Accession Number | O94916 |
| Common Name | Human NFAT5 (aa 1439-1456) |
| Concentration | 0.5 mg/mL |
| Content And Storage | -20°C. Avoid freeze-thaw cycles. |
| Description | Synthetic Peptide |
| Format | Lyophilized |
| Formulation | peptide with no preservative |
| Show More |