Learn More
Biomatik Corporation Recombinant Human Neutrophil elastase (ELANE), 100UG
Catalog No. 501504550
Encompass
Recombinant Human Neutrophil elastase (ELANE) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: ELANE. Target Synonyms: Bone marrow serine protease; Elastase-2; Human leukocyte elastase; HLEMedullasin; PMN elastase. Accession Number: P08246. Expression Region: 30~267aa. Tag Info: N-Terminal Gst-Tagged. Theoretical MW: 52.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 501504550 Supplier Biomatik Corporation Supplier No. RPC20035
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Human Neutrophil elastase (ELANE) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: ELANE. Target Synonyms: Bone marrow serine protease; Elastase-2; Human leukocyte elastase; HLEMedullasin; PMN elastase. Accession Number: P08246. Expression Region: 30~267aa. Tag Info: N-Terminal Gst-Tagged. Theoretical MW: 52.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.