missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human/Mouse/Rat SERCA3 ATPase (aa 29-39) Synthetic Peptide

Catalog No. PEP015
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP015 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP015 Supplier Invitrogen™ Supplier No. PEP015

Synthetic Peptide

Mouse, Rat, Human SERCA3 (residues 29-39) Synthetic Peptide, PEP-015, from Invitrogen.

ATP dependent calcium pumps are responsible in part for the maintenance of low cytoplasmic free calcium concentrations. The ATP pumps that reside in intracellular organelles are encoded by a family of structurally related enzymes, termed the sarcoplasmic or endoplasmic reticulum calcium ATPases (SERCA). The SERCA2 gene is subject to tissue dependent processing which is responsible for the generation of SERCA2a muscle-specific isoform expressed in type I (slow) skeletal, cardiac and smooth muscle and the SERCA2b isoform expressed in all cell types. SERCA3 has been found to co-express with SERCA2b in platelets, mast cells, lymphoid cells, and epithelial cells. Due to this pattern of expression it is referred to as the non-muscle form of SERCA.

Specifications

Gene ID (Entrez) 25391, 489, 53313
Species Human, Mouse, Rat
Purification Method purified
Accession Number P18596, Q64518, Q93084
Common Name Human/Mouse/Rat SERCA3 ATPase (aa 29-39)
Concentration 0.5 mg/mL
Content And Storage -20°C. Avoid freeze-thaw cycles.
Description Synthetic Peptide
Format Lyophilized
Formulation peptide with no preservative
For Use With (Application) Control
Gene Symbol ATP2A3
Lyophilized Lyophilized
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Sequence V - T - D - A - R - E - R - Y - G - P - N
Show More Show Less