missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Human Mesenchymal Stem Cells (Bone Marrow)

Catalog No. SCC034
Change view
Click to view available options
Quantity:
1 x 106cells/vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCC034 1 x 106cells/vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCC034 Supplier MilliporeSigma™ Supplier No. SCC034

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For differentiation to multiple lineages including adipocytes, osteoblasts and chondrocytes

  • Defined as self-renewing population of adherent, bone-marrow-derived multipotent progenitor cells
  • Readily differentiates into lineage-specific cells that form bone, cartilage, fat, tendon, and muscle tissues
  • Isolated from normal human bone marrow
  • Cryopreserved at P2 to ensure optimal phenotype
  • Highest viability and plating efficiency
  • Cells proliferate as adherent cell monolayer

Specifications

Cell Type Human Mesenchymal Stem Cells
For Use With (Application) Cell Differentiation, Cell Culture, Stem Cell Culture
Quantity 1 x 106cells/vial

For Research Use Only