missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Human Lower Jaw Model

Catalog No. S27458
Change view
Click to view available options
Dimensions (L x W x H):
4.25 x 4.5 x 3.25 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
S27458 4.25 x 4.5 x 3.25 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27458 Supplier Eisco™ Supplier No. AMCH1046AS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes 16 extractable teeth and features a partial section of the jaw that can be removed for further study

  • Anatomically correct-model
  • Natural size and color
  • Realistic details, textures, and bony landmarks
  • Ideal for educational and medical demonstrations
  • Made of durable, washable PVC plastic to withstand years of student and instructor use
  • Use for individual study or as replacement for missing/damaged pieces to a full skeletal model set

Specifications

Color Natural
Dimensions (L x W x H) 4.25 x 4.5 x 3.25 in.
For Use With (Application) Educational and Medical Demonstrations
Includes 16 Extractable Teeth
Material PVC
Product Type Anatomical Model, Human