missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Kinesin 5A (aa 1007-1027) Synthetic Peptide

Catalog No. PEP197
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP197 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP197 Supplier Invitrogen™ Supplier No. PEP197

Synthetic Peptide

Human Kinesin 5A (1007-1027) Synthetic Peptide, PEP-197, from Invitrogen.

This gene encodes a member of the kinesin family of proteins. Members of this family are part of a multisubunit complex that functions as a microtubule motor in intracellular organelle transport. Mutations in this gene cause autosomal dominant spastic paraplegia 10.

Specifications

Gene ID (Entrez) 3798
Species Human
Purification Method purified
Accession Number Q12840
Common Name Human Kinesin 5 A (aa 1007-1027)
Concentration 0.5 mg/mL
Content And Storage -20°C. Avoid freeze-thaw cycles.
Description Synthetic Peptide
Format Lyophilized
Formulation peptide with no preservative
For Use With (Application) Control
Gene Symbol KIF5A
Lyophilized Lyophilized
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Sequence C(1007) G Y E A E D Q A K L F P L H Q E T A A S(1027)
Show More Show Less