missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human KIAA1024 (aa 537-668) Control Fragment Recombinant Protein

Catalog No. RP90443
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP90443 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP90443 Supplier Invitrogen™ Supplier No. RP90443
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (89%), Rat (89%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-52839 (PA5-52839. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Specifications

Accession Number Q9UPX6
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 23251
Name Human KIAA1024 (aa 537-668) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AF529169; cDNA sequence AF529169; DD1; KIAA1024; Major intrinsically disordered Notch2-binding receptor 1; Membrane integral NOTCH2-associated receptor 1; MINAR1; Protein DD1; Ubiquitination and mTOR signaling protein; Ubtor; UPF0258 protein KIAA1024
Common Name KIAA1024
Gene Symbol MINAR1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence IQSSCYNSTGSLSQLHKSDCDSSPEHNLTKIANGVPNSKGDKGNRPENTHHSEEELKTSVCKLVLRIGEIERKLESLSGVRDEISQVLGKLNKLDQKMQQPEKVSVQIDLNSLTSEGPSDDSASPRMFHAHS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less