missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human INO80C (aa 36-104) Control Fragment Recombinant Protein

Catalog No. RP98245
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP98245 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP98245 Supplier Invitrogen™ Supplier No. RP98245
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (86%), Rat (86%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61869 (PA5-61869. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

INO80C is a proposed core component of the chromatin remodeling INO80 complex which is involved in transcriptional regulation, DNA replication and probably DNA repair.

Specifications

Accession Number Q6PI98
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 125476
Name Human INO80C (aa 36-104) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias C18orf37; hIes6; IES6; IES6 homolog; INO80 complex subunit C; INO80C
Common Name INO80C
Gene Symbol INO80C
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GYGASKKKKASASSFAQGISMEAMSENKMVPSEFSTGPVEKAAKPLPFKDPNFVHSGHGGAVAGKKNRT
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less