Learn More
ACROBiosystems Inc Biotinylated Human IL-28 R alpha Protein, His,Avitag™
Catalog No. 502719274
Encompass
IL-28 R alpha (Interleukin-28 receptor subunit alpha) also known as IFNLR1 (IFN-lambda receptor 1), it heterodimerizes with another subunit, IL-10RB, to form a type II cytokine receptor ,which binds IL-28A, IL-28B and IL-29. These ligands, which are distantly related to class I in terferons, are activated upon viral infection and trigger the JAK-STAT signaling pathway to exert antiviral activity. The ligand/receptor complex stimulate the activation of the JAK/STAT signaling pathway leading to the expression of IFN-stimulated genes (ISG), which contribute to the antiviral state. Determines the cell type specificity of the lambda interferon action. Shows a more restricted pattern of expression in the epithelial tissues thereby limiting responses to lambda interferons primarily to epithelial cells of the respiratory, gastrointestinal, and reproductive tracts.
Catalog No. 502719274 Supplier ACROBiosystems Inc Supplier No. ILRH82E4200UG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
IL-28 R alpha (Interleukin-28 receptor subunit alpha) also known as IFNLR1 (IFN-lambda receptor 1), it heterodimerizes with another subunit, IL-10RB, to form a type II cytokine receptor ,which binds IL-28A, IL-28B and IL-29. These ligands, which are distantly related to class I in terferons, are activated upon viral infection and trigger the JAK-STAT signaling pathway to exert antiviral activity. The ligand/receptor complex stimulate the activation of the JAK/STAT signaling pathway leading to the expression of IFN-stimulated genes (ISG), which contribute to the antiviral state. Determines the cell type specificity of the lambda interferon action. Shows a more restricted pattern of expression in the epithelial tissues thereby limiting responses to lambda interferons primarily to epithelial cells of the respiratory, gastrointestinal, and reproductive tracts.