missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IL-17A Recombinant Protein

Catalog No. PHC0174
Encompass
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PHC0174 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PHC0174 Supplier Gibco™ Supplier No. PHC0174
Only null left

Recombinant Protein

Carrier-Free Reconstitution: We recommend that the vial be briefly centrifuged prior to opening to bring the contents to the bottom. This lyophilized protein should be reconstituted in sterile, distilled water or buffer to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2°C to 8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein, such as 0.1% BSA and store in working aliquots at -20°C to -80°C. Storage: The lyophilized protein is stable at room temperature for 1 month, but should be kept at -20°C for long term storage. Working aliquots stored with a carrier protein are stable for at least 10 months at -20°C to -80°C. Avoid repeated freeze/thaw cycles.

Interleukin-17A (IL-17A, CTLA-8) is a CD4+ T cell-derived cytokine that promotes inflammatory responses in cell lines and is elevated in rheumatoid arthritis, asthma, multiple sclerosis, psoriasis, and transplant rejection. IL-17A is a 32 kDa long, disulfide-linked homodimer consisting of 136 amino acids that is a member of a six-species family of proteins (IL-17A-17F) and signals through the IL-17 receptor (IL-17R/CDw217). High levels of IL-17A homodimer are produced by activated peripheral blood CD4+ T-cells, and IL-17A also enhances expression of the intracellular adhesion molecule-1 (ICAM-1) in human fibroblasts. In particular, human IL-17A also stimulates epithelial, endothelial, or fibroblastic cells to secrete IL-6, IL-8, G-CSF, and PGE2. In the presence of human IL-17A, fibroblasts can sustain the proliferation of CD34+ hematopoietic progenitors and induce maturation into neutrophils. Mouse, rat, and human IL-17A can induce IL-6 secretion in mouse stromal cells, indicating that all homologs can recognize the mouse receptor. IL-17A regulates the activities of NF-kappa B and mitogen-activated protein kinases, stimulates the expression of IL-6 and cyclooxygenase-2 (PTGS2/COX-2), and enhances the production of nitric oxide (NO).

Specifications

Accession Number Q16552
For Use With (Application) Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 3605
Molecular Weight (g/mol) 31 kDa
Name Human IL-17A
Purification Method Purified
Quantity 25 μg
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 ng/μg
Gene Alias Ctla8; CTLA-8; cytokine; cytotoxic T-lymphocyte-associated antigen 8; cytotoxic T-lymphocyte-associated protein 8; il 17; Il17; IL-17; IL17A; IL-17a; ILN; Interleukin; interleukin 17; interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8); interleukin 17 precursor; interleukin 17A; interleukin-17; Interleukin17A; interleukin-17A; R-IL-17-A
Common Name IL-17A
Gene Symbol IL17A
Biological Activity ED50 = 0.5 - 5.0 ng/mL; determined by the dose-dependent production of GRO-alpha by HT-29 cells.
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Sequence Human IL-17A recombinant protein is homodimeric containing two 137 amino acid subunits
Content And Storage -20°C
Classification Carrier-Free
Activity ED50 = 0.5 - 5.0 ng/mL; determined by the dose-dependent production of GRO-alpha by HT-29 cells.
Shipping Condition Approved for shipment on Wet or Dry Ice
Expression System E. coli
Protein Family Cytokines & Receptors
Protein Form Recombinant, Ligand
Form Lyophilized
Protein Subtype Interleukins
Research Category Immunology, Neurobiology, Inflammation, Angiogenesis, Signal Transduction
Product Line Gibco
Purity or Quality Grade ≥98% by SDS-PAGE and HPLC
Show More Show Less