missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco™ Human Humerus Bone Model
Life-size human humerus bone model provides an incredible amount of realism and detail for examining the human skeleton
Supplier: Eisco™ AMCH1035AS
Description
- Life-size, anatomically correct replica
- Realistic details, textures, and bony landmarks
- Made of durable and sturdy PVC plastic
- Ideal for both student and patient educational demonstrations
- Washable
Chemical Identifiers
| Examining The Human Skeleton | |
| PVC | |
| Anatomical Model, Human |
| 11.5 in. | |
| 2.5 in. |
Specifications
| Examining The Human Skeleton | |
| 11.5 in. | |
| 2.5 in. |
| Left Side | |
| PVC | |
| Anatomical Model, Human |