Learn More
ACROBiosystems Inc ACROBiosystems Human GCGR / Glucagon receptor Protein, His Tag(MALSverified)
Catalog No. 502106358
Encompass
Recombinant Protein;50UG;Human GCGR, His Tag (GCR-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Lys 136 (Accession # P47871-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 14.9 kDa. The protein migrates as 28-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 502106358 Supplier ACROBiosystems Inc Supplier No. GCRH52H350UG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;50UG;Human GCGR, His Tag (GCR-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Lys 136 (Accession # P47871-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 14.9 kDa. The protein migrates as 28-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.