missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Estrogen Receptor alpha (aa 247-261) Synthetic Peptide
Synthetic Peptide
Supplier: Invitrogen™ PEP013
Description
Estrogen Receptor A (ERa) (residues 247-261) Synthetic Peptide, PEP-013, from Invitrogen.
Estrogen Receptors (ER) are members of the steroid/thyroid hormone receptor superfamily of nuclear receptors. The estrogen receptor is a ligand-activated transcription factor, that when bound to estrogen hormone, induces a conformational change that allows dimerization and binding to estrogen response elements (ERE) in DNA. When bound to EREs, ER can positively or negatively regulate gene transcription through the recruitment of coactivator or corepressor proteins. There are two different forms of the estrogen receptor, alpha and beta, encoded by separate genes (ESR1 and ESR2, respectively). Due to alternative RNA splicing, at least 4 estrogen receptor-alpha isoforms are known to exist (Isoform 1 (66 kDa), Isoform 2 (53 kDa), Isoform 3 (47 kDa), Isoform 4 (35 kDa)). Estrogen receptors are widely expressed in different tissue types and are essential for sexual development and reproductive function. They also play a role in other tissues such as bone. Estrogen receptors are involved in pathological processes including breast cancer, endometrial cancer, and osteoporosis.Specifications
| 2099 | |
| purified | |
| Human Estrogen Receptor alpha (aa 247-261) | |
| -20°C. Avoid freeze-thaw cycles. | |
| Lyophilized | |
| Control | |
| Lyophilized | |
| Peptide | |
| RUO - research use only |
| Human | |
| P03372 | |
| 0.5 mg/mL | |
| Synthetic Peptide | |
| peptide with no preservative | |
| ESR1 | |
| No preservative | |
| 50 μg | |
| E - V - G - M - M - K - G - G - I - R - K - D - R - R - G |