missing translation for 'onlineSavingsMsg'
Learn More

Human Epidermal Melanocytes, neonatal, darkly pigmented donor, (HEMn-DP), Proliferating Culture, Prepared in Medium 254 and HMGS

Catalog No. C20225PA
Encompass
Change view
Click to view available options
Quantity:
3 flasks
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20225PA 3 flasks
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C20225PA Supplier Thermo Fisher Scientific Supplier No. C20225PA

Tertiary cultures established from cryopreserved cells, grown to approximately 50% confluence in T-25 flasks.

Specifications

Content And Storage Upon receipt, follow the instructions in the manual.
Age Newborn (14 days or less)
Cell Type Melanocyte Cells
Culture Type Adherent Cell Culture
Form Proliferating Culture
Species Human
Donor Attributes Darkly Pigmented
Donor Source Single Donor
Product Line Cascade Biologics
Product Type Human Epidermal Melanocyte
Quantity 3 flasks
Shipping Condition Room Temperature
Tested For Hepatitis B (HBV), Hepatitis C (HCV), Sterility Testing (Bacteria & Fungi), Mycoplasma, Growth Performance Assay, HIV-1
Show More Show Less
Do you have ethnicity information for your human epidermal melanocyte donor lines?

We do provide age/sex/pigmentation/ethnicity on the Certificate of Analysis (COA).

For Research Use Only. Not for use in diagnostic procedures.