missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human Epidermal Melanocytes, adult, lightly pigmented donor, (HEMa-LP)

Catalog No. C0245C
Encompass
Change view
Click to view available options
Quantity:
1 vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0245C 1 vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C0245C Supplier Gibco™ Supplier No. C0245C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Primary human epidermal melanocytes isolated from adult skin, cryopreserved at the end of the secondary culture.

Primary human epidermal melanocytes isolated from adult skin, cryopreserved at the end of the secondary culture. These cells are isolated from lightly pigmented tissue.

  • Viability at least 70%
  • ≥500,000 viable cells/vial
  • Capable of at least 16 population doublings

Tested for:

  • Mycoplasma: Not Detected
  • Hepatitis B: Not Detected
  • Hepatitis C: Not Detected
  • HIV-1: Not Detected
  • Bacteria, yeast, and other fungi: Not Detected
  • Positive for Mel-5 antigen in the fourth culture after thawing

Specifications

Content And Storage Liquid nitrogen temperature
Age Adult
Cell Type Melanocyte Cells
Culture Type Adherent Cell Culture
Form Cryopreserved
Species Human
Donor Attributes Lightly Pigmented
Donor Source Single Donor
No. of Cells ≥500,000
Product Line Cascade Biologics
Product Type Human Epidermal Melanocyte
Quantity 1 vial
Tested For Hepatitis B (HBV), Hepatitis C (HCV), Sterility Testing (Bacteria and Fungi), Mycoplasma, Growth Performance Assay, HIV-1
Show More Show Less
Do you have ethnicity information for your human epidermal melanocyte donor lines?

We do provide age/sex/pigmentation/ethnicity on the Certificate of Analysis (COA).

What media do you use during the first passage of primary cells?

We use various media and supplements for the various primary cells. Details can be found here (https://www.thermofisher.com/us/en/home/life-science/cell-culture/primary-cell-culture.html).

How is the population doubling calculated for primary cells?

Cumulative population doublings are calculated from the seeding density and harvesting density from each culture level.

What are typical primary cell yields from tissue?

It varies a lot depending on the age of donor, size of tissues, locations, and conditions arrived to our facility. Sometime a small amount of tissue gives us many cells and sometimes a large amount of tissue gives us a very small lot. Typically, we can get 30-300 vials.

Are primary cells isolated from tissue using enzymatic digestion?

Yes, our primary cells are isolated from tissue using enzymatic digestion.

What are primary cells?

Primary cells are cells taken directly from living tissue (e.g., biopsy material) and established for growth in vitro. These cells have undergone very few population doublings and are therefore more representative of the main functional component of the tissue from which they are derived in comparison to continuous (tumor or artificially immortalized) cell lines, making primary cells a more representative model for the in vivo state.

Primary cells from different species may be used, allowing you to highlight potential differences between humans and preclinical test species. Before in vivo studies, mouse or rat cells can be used to refine doses and reduce the number of animals required for preclinical toxicology. Human cells can be used to determine the accuracy of extrapolating human data from an animal model.


For Research Use Only. Not for use in diagnostic procedures.