Biotinylated Human EphA2 Protein HisAvitag (EP2-H82E4) is expressed from human 293 cells (HEK293). It contains AA Ala 24 - Asn 534 (Accession # P29317).
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Biotinylated Human EphA2 Protein HisAvitag (EP2-H82E4) is expressed from human 293 cells (HEK293). It contains AA Ala 24 - Asn 534 (Accession # P29317).