Learn More
Eisco™ Human Ear Labyrinth Model
Catalog No. S24453
Change view
Click to view available options
:
Human Ear Labyrinth Model
Catalog No.
S24453 Human Ear Labyrinth Model
1 options
Catalog No. S24453 Supplier Eisco™ Supplier No. AM220AS
Only null left
Highly detailed model for study of inner ear.
  • Showcases major structures like superior semicircular canal, saccule and utricle
  • Cochlea dissected along longitudinal axis
  • Rod-mounted for 360-degree view
  • Includes key card with 23 call-outs
Specifications
For Use With (Application) study of inner ear
Includes key card with 23 call-outs
Material Plastic
Mounting Rod-mounted for 360-degree view
Size X-Large
Product Type Model, Human Ear Labyrinth