ActiveMax Human DLL4 uBeads premium grade (for cells) are uniform superparamagnetic beads of 5.5 m with streptavidin on its surface and coupled with biotinylated Human DLL4 Protein expressed from human 293 cells (HEK293) and contains AA Ser 27 - Pro 524 (Accession # NP_061947.1 ).