Learn More
ACROBiosystems Inc PE-Labeled Human DLL3 Protein, His Tag (Site-specific conjugation)
Catalog No. 502331075
Encompass
PE-Labeled Human DLL3 Protein, His Tag (DL3-HP2H7) is produced via site-specific conjugation of PE to Human DLL3 Protein, His Tag under optimal conditions with a proprietary technology. Human DLL3 Protein, His Tag is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Leu 492 (Accession # Q9NYJ7-1).
Catalog No. 502331075 Supplier ACROBiosystems Inc Supplier No. DL3HP2H725TESTS
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
PE-Labeled Human DLL3 Protein, His Tag (DL3-HP2H7) is produced via site-specific conjugation of PE to Human DLL3 Protein, His Tag under optimal conditions with a proprietary technology. Human DLL3 Protein, His Tag is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Leu 492 (Accession # Q9NYJ7-1).