missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Human Digestive System Hand-Painted Model

Catalog No. S13515
Change view
Click to view available options
Two parts
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S13515 Two parts
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S13515 Supplier Eisco™ Supplier No. AM0094

Detailed depiction of external and internal parts of the digestive system.

  • Shows nose, mouth cavity and pharynx, esophagus, gastrointestinal tract, liver with gall bladder, pancreas and spleen
  • Duodenum, cecum and rectum are opened
  • Transverse colon removable
  • Mounted on base
  • Supplied with English key card

Specifications

Color Hand Painted
Dimensions (L x W x H) 45 x 20 x 6 cm
For Use With (Application) Depicts external and internal parts of the digestive system
Includes model, key card
Material ABS Plastic
Mounting Base
Weight (Metric) 740 g
Product Type Digestive System Model