Learn More
ACROBiosystems Inc Human CXCR3 Full Length Protein (VLP)
Catalog No. 502331805
Encompass
Human CXCR3 Full Length Protein-VLP (CX3-H52P4) is expressed from human 293 cells (HEK293). It contains AA Val 2 - Leu 368 (Accession # P49682-1).
Catalog No. 502331805 Supplier ACROBiosystems Inc Supplier No. CX3H52P420UG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Human CXCR3 Full Length Protein-VLP (CX3-H52P4) is expressed from human 293 cells (HEK293). It contains AA Val 2 - Leu 368 (Accession # P49682-1).