missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CREB (aa 123-136) Synthetic Peptide

Catalog No. PEP143
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP143 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP143 Supplier Invitrogen™ Supplier No. PEP143

Synthetic Peptide

Human CREB (residues 123-136) Synthetic Peptide, PEP-143, from Invitrogen.

CREB (Cyclic AMP response element binding protein) is a 43 kDa basic/leucine zipper transcription factor that binds the cyclic AMP response element (CRE) and activates transcription in response to a variety of extracellular signals including neurotransmitters, hormones, membrane depolarization, and growth and neurotrophic factors. Activation of CREB is dependent upon the phosphorylation of serine 133. Phosphorylation of CREB occurs via p44/42 MAP kinase and p90RSK, and via p38 MAP kinase and MSK1. Although CREB will bind DNA independent of its phosphorylation state, only the phosphorylated form is competent as a transcription factor. CREB binding protein (CBP), a transcriptional coactivator that directly interacts with CREB, binds to CREB in the region of serine 133. CREB is involved in different cellular processes including the synchronization of circadian rhythmicity, differentiation of adipose cells, and learning and memory. In humans, the gene is located on the q arm of chromosome 2. Diseases associated with CREB dysfunction include Alzheimer’s Disease, histiocytoma and soft tissue melanoma. PEP-143 is a 14 amino acid synthetic phospho-peptide whose sequence corresponds to residues 123-136 of human CREB with serine 133 being phosphorylated. The sequence of this peptide is (amino to carboxy terminus): K (123)-R-R-E-I-L-S-R-R-P-S (p)-Y-R-K (136)This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and human CREB, catalog &Num; PA1-851B. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∽70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 mL of distilled water.

Specifications

Gene ID (Entrez) 1385
Species Human
Purification Method purified
Accession Number P16220
Common Name Human CREB (aa 123-136)
Concentration 0.5 mg/mL
Content And Storage -20°C. Avoid freeze-thaw cycles.
Description Synthetic Peptide
Format Lyophilized
Formulation peptide with no preservative
For Use With (Application) Control
Gene Symbol CREB1
Lyophilized Yes
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Sequence K(123)-R-R-E-I-L-S-R-R-P-S(p)-Y-R-K(136)
Show More Show Less