Learn More
ACROBiosystems Inc Biotinylated Human Complement C5 Protein, His,Avitag™
Catalog No. 502331310
Encompass
Biotinylated Human Complement C5, His,Avitag (CO5-H82E9) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Cys 1676 (Accession # P01031-1).
Catalog No. 502331310 Supplier ACROBiosystems Inc Supplier No. CO5H82E925UG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Biotinylated Human Complement C5, His,Avitag (CO5-H82E9) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Cys 1676 (Accession # P01031-1).