missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Human Circulatory System Model

Catalog No. S13513
Change view
Click to view available options
Grade:
9 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S13513 9 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S13513 Supplier Eisco™ Supplier No. AM0092

Details entire blood circulation system in veins and arteries.

  • Hand painted
  • Mounted on base
  • Supplied with English key card

Specifications

Color Brown
Dimensions (L x W x H) 45 x 20 x 6 cm
For Use With (Application) Details entire blood circulation system in veins and arteries
Grade 9 to 12
Includes Model, English key card
Material ABS Plastic
Mounting Base
Quantity 1
Gender Male, Female
Product Type Circulatory System Model