Learn More
Acrobiosystems FITC-Labeled Human CD19 (20-291), His Tag (CD9-HF2H3) is produced via conjugation of FITC to Human CD19 (20-291), with a new generation site-specific technology under Star Staining labeling platform. AA Pro 20 - Lys 291 ( # P15391-1). DFS Item
Supplier:  ACROBiosystems Inc CD9HF2H325UG
B-lymphocyte antigen CD19 is also known as CD19 (Cluster of Differentiation 19), is a single-pass type I membrane protein which contains twoIg-like C2-type (immunoglobulin-like) domains. CD19 is expressed on follicular dendritic cells and B cells. In fact, it is present on B cells from earliest recognizable B-lineage cells during development to B-cell blasts but is lost on maturation to plasma cells. It primarily acts as a B cell co-receptor in conjunction with CD21 and CD81. Upon activation, the cytoplasmic tail of CD19 becomes phosphorylated, which leads to binding by Src-family kinases and recruitment of PI-3 kinase. As on T cells, several surface molecules form the antigen receptor and form a complex on B lymphocytes. The (almost) B cell-specific CD19 phosphoglycoprotein is one of these molecules. The others are CD21 and CD81. These surface immunoglobulin (sIg)-associated molecules facilitate signal transduction.
Catalog No. 502231313
Only null left
Explore available promotions