ACROBiosystems Inc Alexa Fluor 647-Labeled Human CD19 (20-291) Protein, His TagStar Staining
Catalog No. 502331280
Alexa Fluor 647-Labeled Human CD19 (20-291), His Tag (CD9-HA2H6) is produced via conjugation of AF647 to Human CD19 (20-291), His Tag with a new generation site-specific technology under Star Staining labeling platform. Human CD19 (20-291), His Tag is expressed from human 293 cells (HEK293). It contains AA Pro 20 - Lys 291 (Accession # P15391-1).
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Alexa Fluor 647-Labeled Human CD19 (20-291), His Tag (CD9-HA2H6) is produced via conjugation of AF647 to Human CD19 (20-291), His Tag with a new generation site-specific technology under Star Staining labeling platform. Human CD19 (20-291), His Tag is expressed from human 293 cells (HEK293). It contains AA Pro 20 - Lys 291 (Accession # P15391-1).