missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CARD9 Synthetic Peptide

Catalog No. PEP0115
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0115 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0115 Supplier Invitrogen™ Supplier No. PEP0115

Synthetic Peptide

This peptide corresponds to 16 amino acids of human CARD9. PEP-0115 can be used as a blocking peptide with polyclonal antibody PA5-19993.

The protein encoded by this gene is a member of the CARD protein family, which is defined by the presence of a characteristic caspase-associated recruitment domain (CARD). CARD is a protein interaction domain known to participate in activation or suppression of CARD containing members of the caspase family, and thus plays an important regulatory role in cell apoptosis. This protein was identified by its selective association with the CARD domain of BCL10, a postive regulator of apoptosis and NF-kappaB activation, and is thought to function as a molecular scaffold for the assembly of a BCL10 signaling complex that activates NF-kappaB. Several alternatively spliced transcript variants have been observed, but their full-length nature is not clearly defined.

Specifications

Species Human
Purification Method purified
Accession Number Q9H257
Common Name CARD9
Concentration 200 μg/mL
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
Gene Alias CANDF2; card 9; Card9; caspase recruitment domain family member 9; caspase recruitment domain family, member 9; caspase recruitment domain protein 9; caspase recruitment domain-containing protein 9; Gm782; hCARD9; rCARD9; RP11-413M3.9
Gene Symbol CARD9
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Show More Show Less