missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human C1orf74 (aa 176-255) Control Fragment Recombinant Protein

Catalog No. RP94407
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP94407 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP94407 Supplier Invitrogen™ Supplier No. RP94407
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (79%), Rat (79%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55879 (PA5-55879. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Specifications

Accession Number Q96LT6
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 148304
Name Human C1orf74 (aa 176-255) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias C1orf74; chromosome 1 open reading frame 74; UPF0739 protein C1orf74; up-regulated in lung cancer 4; URLC4
Common Name C1orf74
Gene Symbol C1orf74
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LLGYPVPYTFHLNQGDDNCLALTPLRVFTARISWLLGQPPILLYSFSVPESLFPGLRDILNTWEKDLRTRFRTQNDFADL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less