missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Brain Reference RNA for QuantiGene Assays

Catalog No. 361055672
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
361055672 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 361055672 Supplier Invitrogen™ Supplier No. QS0611

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Human Brain Total RNA is a total RNA sample extracted from neural tissue for use as a positive control in QuantiGene assays.

Human Brain Total RNA is a total RNA sample extracted from neural tissue for use as a positive control in QuantiGene assays.

Spécifications

Species Human
Sample Type RNA
For Use With (Application) Nucleic Acid Quantitation
Product Type Reference RNA
Quantity 10 μg
Sample Source Pooled, Normal Tissue
Shipping Condition Dry Ice