Learn More
ACROBiosystems Inc Alexa Fluor 647-Labeled Human BCMA / TNFRSF17 Protein, His TagStar Staining
Catalog No. 502331882
Encompass
Alexa Fluor 647-Labeled Human BCMA, His Tag (BCA-HA2H9) is produced via conjugation of AF647 to Human BCMA, His Tag with a new generation site-specific technology under Star Staining labeling platform. Human BCMA, His Tag is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 54 (Accession # Q02223-1).
Catalog No. 502331882 Supplier ACROBiosystems Inc Supplier No. BCAHA2H9200UG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Alexa Fluor 647-Labeled Human BCMA, His Tag (BCA-HA2H9) is produced via conjugation of AF647 to Human BCMA, His Tag with a new generation site-specific technology under Star Staining labeling platform. Human BCMA, His Tag is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 54 (Accession # Q02223-1).