missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Inc Biotinylated Human B7-H7 (HHLA2) Protein, Fc-Avitag - 25 g

Catalog No. 502370253
Encompass

Biotinylated Human B7-H7 (HHLA2) is a recombinant Fc-tagged extracellular domain produced in human 293 (HEK293) cells and supplied lyophilized with trehalose. It is enzymatically biotinylated for site-specific binding assays and has been MALS-verified for quality, making it suitable for bioactivity ELISA applications targeting CD28H.

  • Recombinant extracellular domain of human B7-H7 (HHLA2) expressed in HEK293 cells.
  • Human IgG1 Fc tag for dimerization and detection.
  • Site-specific enzymatic biotinylation at a single lysine residue.
  • Purity >95% by SDS-PAGE and >90% by SEC-MALS.
  • Calculated MW 65.1 kDa; apparent 70-90 kDa on reducing SDS-PAGE due to glycosylation.
  • Lyophilized formulation with trehalose; formulated in Tris/glycine/arginine/NaCl buffer.
  • Validated for bioactivity ELISA binding to CD28H with QC-tested linear range in the low ng/mL.

Catalog No. 502370253 Supplier ACROBiosystems Inc Supplier No. B77H82F325UG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Biotinylated Human B7-H7 (HHLA2) is a recombinant Fc-tagged extracellular domain produced in human 293 (HEK293) cells and supplied lyophilized with trehalose. It is enzymatically biotinylated for site-specific binding assays and has been MALS-verified for quality, making it suitable for bioactivity ELISA applications targeting CD28H.

  • Recombinant extracellular domain of human B7-H7 (HHLA2) expressed in HEK293 cells.
  • Human IgG1 Fc tag for dimerization and detection.
  • Site-specific enzymatic biotinylation at a single lysine residue.
  • Purity >95% by SDS-PAGE and >90% by SEC-MALS.
  • Calculated MW 65.1 kDa; apparent 70-90 kDa on reducing SDS-PAGE due to glycosylation.
  • Lyophilized formulation with trehalose; formulated in Tris/glycine/arginine/NaCl buffer.
  • Validated for bioactivity ELISA binding to CD28H with QC-tested linear range in the low ng/mL.