missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ARMET Synthetic Peptide

Catalog No. PEP0549
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0549 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0549 Supplier Invitrogen™ Supplier No. PEP0549

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Synthetic Peptide

This peptide corresponds to 12 amino acids near the amino terminus of human MANF. PEP-0549 can be used as a blocking peptide with polyclonal antibody PA5-20432.

The protein encoded by this gene is localized in the endoplasmic reticulum and golgi, and is also secreted. Reducing expression of this gene increases susceptibility to ER stress-induced death and promotes cell proliferation. The protein was initially thought to be longer at the N-terminus and to contain an arginine-rich region but transcribed evidence indicates a smaller open reading frame that does not encode the arginine tract. The presence of polymorphisms in the arginine-rich region, including a specific mutation that changes the previously numbered codon 50 from ATG to AGG, have been reported in a variety of solid tumors; however, these polymorphisms were later shown to exist in normal tissues and are thus not tumor-related.

Specifications

Gene ID (Entrez) 7873
Species Human
Purification Method purified
Accession Number P55145
Common Name ARMET
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias 3230402M22Rik; AA407711; AA408789; AA673178; Arginine-rich protein; arginine-rich, mutated in early stage tumors; ARMET; ARP; D17914; D18Mgi17; Manf; mesencephalic astrocyte derived neurotrophic factor; mesencephalic astrocyte-derived neurotrophic factor; MGC142148; MGC142150; Protein ARMET
Gene Symbol MANF
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less