missing translation for 'onlineSavingsMsg'
Learn More

Potassium Ferricyanide, Granular, Technical, Spectrum™ Chemical

Catalog No. 18612621
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612621 50 kg Amber Glass Bottle
18612618 500 g Amber Glass Bottle
18612619 2.5 kg Amber Glass Bottle
18612620 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18612621 Supplier Spectrum Chemical Mfg Cor Supplier No. P128650KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13746-66-2

Specifications

CAS Max % 1
Physical Form Granular/Lump
Quantity 50 kg
Molecular Weight (g/mol) 329.25
Grade Technical
Packaging Amber Glass Bottle
Chemical Name or Material Potassium Ferricyanide, Granular

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.