missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human AIF1L (aa 85-150) Control Fragment Recombinant Protein

Catalog No. RP92944
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP92944 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP92944 Supplier Invitrogen™ Supplier No. RP92944
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (98%), Rat (98%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-54226 (PA5-54226. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

AIF1L Actin-binding protein that promotes actin bundling. May neither bind calcium nor depend on calcium for function.

Specifications

Accession Number Q9BQI0
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 83543
Name Human AIF1L (aa 85-150) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AIF1L; allograft inflammatory factor 1 like; allograft inflammatory factor 1-like; C9orf58; IBA2; ionized calcium binding adapter molecule 2; ionized calcium-binding adapter molecule 2; UNQ672/PRO1306
Common Name AIF1L
Gene Symbol AIF1L
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence HLEMKKMISEVTGGVSDTISYRDFVNMMLGKRSAVLKLVMMFEGKANESSPKPVGPPPERDIASLP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less