missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Aggrecan (Interglobular Domain) Recombinant Protein

Catalog No. PIRP77527
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP77527 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP77527 Supplier Invitrogen™ Supplier No. RP77527
Only null left

Recombinant Protein

Aggrecan Interglobular Domain (human) Recombinant Protein, RP-77527, from Invitrogen.

The human aggrecan amino acid sequence T331-G458 is expressed in E. coli with a C-terminal His-tag. The recombinant protein contains cleavage sites for aggrecanases (E373-A374 in aggrecan) and matrix metalloproteinases (N341-F342 in aggrecan). RP-77527 is a purified recombinant Aggrecan Interglobular Domain protein, amino acid sequence T331-G458 expressed in E. coli with a C-terminal His-tag. The recombinant protein contains cleavage sites for aggrecanases (E373-A374 in aggrecan) and matrix metalloproteinases (N341-F342 in aggrecan). RP-77527 may be used as a control with corresponding anti-Aggrecan Interglobular Domain antibodies.

Compliance

in vitro research only.

Specifications

Accession Number P16112
Concentration 0.5mg/mL
For Use With (Application) Control
Formulation TBS with 5mM CaCl2 and no preservative; pH 7.5
Gene ID (Entrez) 176
Name Human Aggrecan (Interglobular Domain)
pH Range 7.5
Quantity 100 μg
Source E. coli
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Common Name Human Aggrecan (Interglobular Domain)
Gene Symbol ACAN
Specific Reactivity Human
Conjugate Unconjugated
Cross Reactivity Hu
Species E. coli
Recombinant Recombinant
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade purified
Show More Show Less