Learn More
Bioivt, Llc Human AB serum 100ml Bottle
Supplier: Valley Biomedical Prod & Srv Inc HUMANABSRMPHI1
Human AB serum derived from apharesis plasma heat inactivated- Steile filtered, mycoplasma and endotoxin tested. The Valley Biomedical heat inactivation protocol is to bring the temperature of the material to 56 deg C and maintain this for 30 minutes. Some precipitation may be evident, which can be removed, if desired, by filtration. 100ml Bottle. Store at -20 deg C.