Learn More
ACROBiosystems Inc ACROBiosystems Human 2B4 / CD244 / SLAMF4 Protein, Fc Tag
Catalog No. 502106199
Encompass
Recombinant Protein;100UG;Human 2B4, Fc Tag (2B4-H5255) is expressed from human 293 cells (HEK293). It contains AA Cys 22 - Arg 221 (Accession # NP_057466).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 48.9 kDa. The protein migrates as 60-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human 2B4, Fc Tag (Cat. No. 2B4-H5255) at 2 g/mL (100 L/well) can bind Human CD48, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. BC1-H5258) with a linear range of 2-16 ng/mL (QC tested).
Catalog No. 502106199 Supplier ACROBiosystems Inc Supplier No. 2B4H5255100UG
Recombinant Protein;100UG;Human 2B4, Fc Tag (2B4-H5255) is expressed from human 293 cells (HEK293). It contains AA Cys 22 - Arg 221 (Accession # NP_057466).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 48.9 kDa. The protein migrates as 60-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human 2B4, Fc Tag (Cat. No. 2B4-H5255) at 2 g/mL (100 L/well) can bind Human CD48, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. BC1-H5258) with a linear range of 2-16 ng/mL (QC tested).