missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human 20S Proteasome Z (aa 263-277) Synthetic Peptide

Catalog No. PEP175
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP175 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP175 Supplier Invitrogen™ Supplier No. PEP175

Synthetic Peptide

Human 20S Proteasome Z (residues 263-277) Synthetic Peptide, PEP-175, from Invitrogen.

The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the proteasome B-type family, also known as the T1B family, that is a 20S core beta subunit in the proteasome. Expression of this catalytic subunit is downregulated by gamma interferon and proteolytic processing is required to generate a mature subunit. This subunit is not present in the immunoproteasome and is replaced by catalytic subunit 2i (proteasome beta 10 subunit).

Specifications

Gene ID (Entrez) 5695
Species Human
Purification Method purified
Accession Number Q99436
Common Name Human 20 S Proteasome Z (aa 263-277)
Concentration 0.5 mg/mL
Content And Storage -20°C. Avoid freeze-thaw cycles.
Description Synthetic Peptide
Format Lyophilized
Formulation peptide with no preservative
For Use With (Application) Control
Gene Symbol PSMB7
Lyophilized Lyophilized
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Sequence C-E(263)-I-E-V-L-E-E-T-V-Q-T-M-D-T-S(NH2)(277)
Show More Show Less