Biotinylated human APRIL (TNFSF13) protein is a recombinant, active trimeric cytokine expressed in HEK293 cells and enzymatically biotinylated. The product is supplied lyophilized in PBS (pH 7.4) with trehalose and is quality-verified by SEC-MALS and SDS-PAGE.
- Expression system: Human 293 cells (HEK293).
- Format: Lyophilized powder in PBS, pH 7.4, with trehalose protectant.
- Biotinylation: Site-specific biotinylation at a single lysine residue.
- Tag: N-terminal polyhistidine tag for purification and detection.
- Purity: >90% by SDS-PAGE and SEC-MALS.
- Molecular weight: Calculated 50.9 kDa; migrates 55-63 kDa on SDS-PAGE.
- Available size: 200 g.