Learn More
Air-Tite HSW Soft-Ject™ Disposable Syringes
Air-Tite HSW Soft-Ject™ Disposable Syringes are high-quality, three-part disposable rubber piston syringes
$158.34 - $158.34
Specifications
| Latex Free | Yes |
|---|---|
| Product Type | Soft-Ject Syringe |
| Disposable | Single-use |
| Sterility | Sterile |
| Catalog Number | Mfr. No. | Volume (Metric) | Connection Type | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Volume (Metric) | Connection Type | Price | Quantity | ||||
|
36-103-1239
|
Air-Tite
R20 |
20 mL | Leur Slip |
Pack of 100 Each for $158.34
|
|
||||
Description
- Lubricated synthetic rubber piston allows for smooth draw while positive stop helps prevent accidental pullout
- Bold graduations are easily read and resist wipe off
- ETO gas sterilized syringes, latex and pyrogen free
- Packaged in individual sterile strip packs
- Made in Germany
Specifications
| Yes | |
| Soft-Ject Syringe | |
| Single-use | |
| Sterile |